Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl, 97%

$230.00 - $230.00

Chemical Identifiers

Chemical Name or Material (S)-(-)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl
CAS 142128-92-5
Molecular Formula C24H22O4
Formula Weight 374.44g/mol
MDL Number MFCD03788935
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3288903

 
alfa aesar™
H32889-03
1g Each for $230.00
Description & Specifications

Specifications

Melting Point 101° to 105°C
Quantity 1g
Chemical Name or Material (S)-(-)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl
CAS 142128-92-5
Assay 97%
Molecular Formula C24H22O4
Formula Weight 374.44g/mol
MDL Number MFCD03788935