Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl-3,3'-diboronic acid pinacol ester, 97%

$266.36 - $266.36

Chemical Identifiers

Chemical Name or Material (S)-(-)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl-3,3'-diboronic acid pinacol ester
CAS 260441-99-4
Molecular Formula C36H44B2O8
Formula Weight 626.36g/mol
MDL Number MFCD16294496
Synonym (S)-2,2'Bis(methoxymethoxy)-3,3'-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,1'-binaphthyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3200503

 
alfa aesar™
H32005-03
1g Each for $266.36
Description & Specifications

Specifications

Melting Point 193° to 195°C
Quantity 1g
Optical Rotation −21.3° (c=1 in DCM)
Chemical Name or Material (S)-(-)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl-3,3'-diboronic acid pinacol ester
CAS 260441-99-4
Assay 97%
Molecular Formula C36H44B2O8
Formula Weight 626.36g/mol
MDL Number MFCD16294496
Synonym (S)-2,2'Bis(methoxymethoxy)-3,3'-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,1'-binaphthyl