missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-2,2'-Bis(diphenylphosphino)-4,4',6,6'-tetramethoxybiphenyl, 97+%

$82.61 - $330.46

Chemical Identifiers

CAS 1365531-76-5
Molecular Formula C40H36O4P2
Molecular Weight (g/mol) 642.672
MDL Number MFCD19443622
InChI Key YSQIPQZYSQJPNL-UHFFFAOYSA-N
Synonym 2'-diphenylphosphanyl-4,4',6,6'-tetramethoxy-1,1'-biphenyl-2-yl diphenylphosphane, r-2,2'-bis diphenylphosphino-4,4',6,6'-tetramethoxybiphenyl, s-2,2'-bis diphenylphosphino-4,4',6,6'-tetramethoxybiphenyl, r-2,2'-bis diphenylphosphino-4,4',6,6'-tetramethoxy-1,1'-biphenyl, s-2,2'-bis diphenylphosphino-4,4',6,6'-tetramethoxy-1,1'-biphenyl, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis diphenylphosphine, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis diphenylphosphine
PubChem CID 18706016
IUPAC Name [2-(2-diphenylphosphanyl-4,6-dimethoxyphenyl)-3,5-dimethoxyphenyl]-diphenylphosphane
SMILES COC1=CC(=C(C(=C1)P(C2=CC=CC=C2)C3=CC=CC=C3)C4=C(C=C(C=C4P(C5=CC=CC=C5)C6=CC=CC=C6)OC)OC)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60069MD View Documents Alfa Aesar™
H60069MD
250mg Each for $82.61
 
AAH6006903 View Documents Alfa Aesar™
H6006903
1g Each for $330.46
 
Specifications

Chemical Identifiers

1365531-76-5
642.672
YSQIPQZYSQJPNL-UHFFFAOYSA-N
18706016
COC1=CC(=C(C(=C1)P(C2=CC=CC=C2)C3=CC=CC=C3)C4=C(C=C(C=C4P(C5=CC=CC=C5)C6=CC=CC=C6)OC)OC)OC
C40H36O4P2
MFCD19443622
2'-diphenylphosphanyl-4,4',6,6'-tetramethoxy-1,1'-biphenyl-2-yl diphenylphosphane, r-2,2'-bis diphenylphosphino-4,4',6,6'-tetramethoxybiphenyl, s-2,2'-bis diphenylphosphino-4,4',6,6'-tetramethoxybiphenyl, r-2,2'-bis diphenylphosphino-4,4',6,6'-tetramethoxy-1,1'-biphenyl, s-2,2'-bis diphenylphosphino-4,4',6,6'-tetramethoxy-1,1'-biphenyl, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis diphenylphosphine, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis diphenylphosphine
[2-(2-diphenylphosphanyl-4,6-dimethoxyphenyl)-3,5-dimethoxyphenyl]-diphenylphosphane

Specifications

250mg
C40H36O4P2
2'-diphenylphosphanyl-4,4',6,6'-tetramethoxy-1,1'-biphenyl-2-yl diphenylphosphane, r-2,2'-bis diphenylphosphino-4,4',6,6'-tetramethoxybiphenyl, s-2,2'-bis diphenylphosphino-4,4',6,6'-tetramethoxybiphenyl, r-2,2'-bis diphenylphosphino-4,4',6,6'-tetramethoxy-1,1'-biphenyl, s-2,2'-bis diphenylphosphino-4,4',6,6'-tetramethoxy-1,1'-biphenyl, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis diphenylphosphine, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis diphenylphosphine
COC1=CC(=C(C(=C1)P(C2=CC=CC=C2)C3=CC=CC=C3)C4=C(C=C(C=C4P(C5=CC=CC=C5)C6=CC=CC=C6)OC)OC)OC
642.672
642.66
(S)-2,2'-Bis(diphenylphosphino)-4,4',6,6'-tetramethoxybiphenyl
1365531-76-5
MFCD19443622
YSQIPQZYSQJPNL-UHFFFAOYSA-N
[2-(2-diphenylphosphanyl-4,6-dimethoxyphenyl)-3,5-dimethoxyphenyl]-diphenylphosphane
18706016
≥97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning