Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 97%

$46.41 - $120.69

Chemical Identifiers

Chemical Name or Material (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
CAS 76189-56-5
Molecular Formula C44H32P2
Formula Weight 622.69
MDL Number MFCD00010805
Beilstein 5321444
Merck Index 14,1223
Synonym (S)-(-)-BINAP
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2387201

  SDS
alfa aesar™
B2387201
0.25g Each for $46.41

AAB2387203

  SDS
alfa aesar™
B2387203
1g Each for $120.69
Description & Specifications

Specifications

Formula Weight 622.69
MDL Number MFCD00010805
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Optical Rotation −230° (c=0.3 in Toluene)
Melting Point 239° to 241°C
Synonym (S)-(-)-BINAP
Merck Index 14,1223
Molecular Formula C44H32P2
Beilstein 5321444
Quantity 0.25g
CAS 76189-56-5
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Chemical Name or Material (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.