missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-2,2'-Bis[bis(3,5-trifluoromethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl, 97+%, Alfa Aesar™

$218.99 - $791.11

Chemical Identifiers

CAS 1365531-85-6
Molecular Formula C48H28F24O4P2
Molecular Weight (g/mol) 1186.657
MDL Number MFCD19443625
InChI Key JDKXXRZDVSJSNQ-UHFFFAOYSA-N
Synonym r-2,2'-bis bis 3,5-trifluoromethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, s-2,2'-bis bis 3,5-trifluoromethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, ar-2,2',4,4'-tetramethoxy-6,6'-bis bis 3,5-bis trifluoromethyl phenyl phosphino biphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 3,5-bis trifluoromethyl phenyl phosphine, r-2,2'-bis bis 3,5-trifluoromethylphenyl phosphino-4,4',6,6'-tetramethoxy-1,1'-biphenyl, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 3,5-bis trifluoromethyl phenyl phosphine, s-2,2'-bis bis 3,5-trifluoromethylphenyl phosphino-4,4',6,6'-tetramethoxy-1,1'-biphenyl, bis 3,5-bis trifluoromethyl phenyl 2'-bis 3,5-bis trifluoromethyl phenyl phosphanyl-4,4',6,6'-tetramethoxy-1,1'-biphenyl-2-yl phosphane
PubChem CID 87111592
IUPAC Name [2-[2-bis[3,5-bis(trifluoromethyl)phenyl]phosphanyl-4,6-dimethoxyphenyl]-3,5-dimethoxyphenyl]-bis[3,5-bis(trifluoromethyl)phenyl]phosphane
SMILES COC1=CC(=C(C(=C1)P(C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)C3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C4=C(C=C(C=C4P(C5=CC(=CC(=C5)C(F)(F)F)C(F)(F)F)C6=CC(=CC(=C6)C(F)(F)F)C(F)(F)F)OC)OC)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60604MD View Documents Alfa Aesar
H60604MD
250mg Each for $218.99
 
AAH6060403 View Documents Alfa Aesar
H6060403
1g Each for $791.11
 
Description

Description

Solubility
Insoluble in water.

Notes
Air sensitive. Store at room temperature.
Specifications

Chemical Identifiers

1365531-85-6
1186.657
JDKXXRZDVSJSNQ-UHFFFAOYSA-N
87111592
COC1=CC(=C(C(=C1)P(C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)C3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C4=C(C=C(C=C4P(C5=CC(=CC(=C5)C(F)(F)F)C(F)(F)F)C6=CC(=CC(=C6)C(F)(F)F)C(F)(F)F)OC)OC)OC
C48H28F24O4P2
MFCD19443625
r-2,2'-bis bis 3,5-trifluoromethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, s-2,2'-bis bis 3,5-trifluoromethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, ar-2,2',4,4'-tetramethoxy-6,6'-bis bis 3,5-bis trifluoromethyl phenyl phosphino biphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 3,5-bis trifluoromethyl phenyl phosphine, r-2,2'-bis bis 3,5-trifluoromethylphenyl phosphino-4,4',6,6'-tetramethoxy-1,1'-biphenyl, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 3,5-bis trifluoromethyl phenyl phosphine, s-2,2'-bis bis 3,5-trifluoromethylphenyl phosphino-4,4',6,6'-tetramethoxy-1,1'-biphenyl, bis 3,5-bis trifluoromethyl phenyl 2'-bis 3,5-bis trifluoromethyl phenyl phosphanyl-4,4',6,6'-tetramethoxy-1,1'-biphenyl-2-yl phosphane
[2-[2-bis[3,5-bis(trifluoromethyl)phenyl]phosphanyl-4,6-dimethoxyphenyl]-3,5-dimethoxyphenyl]-bis[3,5-bis(trifluoromethyl)phenyl]phosphane

Specifications

250mg
C48H28F24O4P2
r-2,2'-bis bis 3,5-trifluoromethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, s-2,2'-bis bis 3,5-trifluoromethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, ar-2,2',4,4'-tetramethoxy-6,6'-bis bis 3,5-bis trifluoromethyl phenyl phosphino biphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 3,5-bis trifluoromethyl phenyl phosphine, r-2,2'-bis bis 3,5-trifluoromethylphenyl phosphino-4,4',6,6'-tetramethoxy-1,1'-biphenyl, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 3,5-bis trifluoromethyl phenyl phosphine, s-2,2'-bis bis 3,5-trifluoromethylphenyl phosphino-4,4',6,6'-tetramethoxy-1,1'-biphenyl, bis 3,5-bis trifluoromethyl phenyl 2'-bis 3,5-bis trifluoromethyl phenyl phosphanyl-4,4',6,6'-tetramethoxy-1,1'-biphenyl-2-yl phosphane
COC1=CC(=C(C(=C1)P(C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)C3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C4=C(C=C(C=C4P(C5=CC(=CC(=C5)C(F)(F)F)C(F)(F)F)C6=CC(=CC(=C6)C(F)(F)F)C(F)(F)F)OC)OC)OC
1186.657
1186.64
(S)-2,2'-Bis[bis(3,5-trifluoromethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl
1365531-85-6
MFCD19443625
JDKXXRZDVSJSNQ-UHFFFAOYSA-N
[2-[2-bis[3,5-bis(trifluoromethyl)phenyl]phosphanyl-4,6-dimethoxyphenyl]-3,5-dimethoxyphenyl]-bis[3,5-bis(trifluoromethyl)phenyl]phosphane
87111592
≥97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning