Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine

Asymmetric hydrogenation

$86.32 - $288.64

Chemical Identifiers

Chemical Name or Material (S)- (-)-2,2',6,6'-Tetramethoxy-4,4'-bis (diphenylphosphino)-3,3'-bipyridine
CAS 362524-23-0
Molecular Formula C38H34N2O4P2
Formula Weight 644.64g/mol
MDL Number MFCD04038734
Synonym (S)-P-Phos
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4460902

 
alfa aesar™
44609-02
0.5g Each for $288.64

AA4460977

 
alfa aesar™
44609-77
0.1g Each for $86.32
Description & Specifications

Specifications

Melting Point 261° to 265°C
Odor Odorless
Quantity 0.5g
Chemical Name or Material (S)- (-)-2,2',6,6'-Tetramethoxy-4,4'-bis (diphenylphosphino)-3,3'-bipyridine
CAS 362524-23-0
Physical Form Powder
Molecular Formula C38H34N2O4P2
Formula Weight 644.64g/mol
MDL Number MFCD04038734
Synonym (S)-P-Phos
Storage Note 1
  • Packaged under Argon

  • Packaged under Argon

Safety and Handling

  • Packaged under Argon