missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-(-)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine, Thermo Scientific Chemicals
Asymmetric hydrogenation
Supplier: Thermo Scientific Chemicals 04461477
Description
(S)-(-)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine is used in Pharmaceutical and industry applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Specifications
(S)- (-)-2,2',6,6'-Tetramethoxy-4,4'-bis (di (3,5-xylyl)phosphino)-3,3'-bipyridine | |
158°C to 162°C | |
MFCD04974235 | |
unii-1a4eo1493y, unii-n2pcw56063, r-4,4'-bis bis 3,5-dimethylphenyl phosphino-2,2',6,6'-tetramethoxy-3,3'-bipyridine, s---2,2',6,6'-tetramethoxy-4,4'-bis di 3,5-xylyl phosphino-3,3'-bipyridine, s-4,4'-bis bis 3,5-dimethylphenyl phosphino-2,2',6,6'-tetramethoxy-3,3'-bipyridine, xyl-p-phos, r, xyl-p-phos, s, r-xyl-p-phos mi, s-xyl-p-phos mi, xyl-p-phos | |
JRTHAKOHBMETRC-UHFFFAOYSA-N | |
[3-[4-bis(3,5-dimethylphenyl)phosphanyl-2,6-dimethoxypyridin-3-yl]-2,6-dimethoxypyridin-4-yl]-bis(3,5-dimethylphenyl)phosphane | |
11182279 | |
Powder |
443347-10-2 | |
C46H50N2O4P2 | |
0.1 g | |
Sparingly soluble in water. | |
CC1=CC(=CC(=C1)P(C2=CC(=CC(=C2)C)C)C3=CC(=NC(=C3C4=C(N=C(C=C4P(C5=CC(=CC(=C5)C)C)C6=CC(=CC(=C6)C)C)OC)OC)OC)OC)C | |
756.864 | |
756.86 |
Chemical Identifiers
443347-10-2 | |
756.864 | |
JRTHAKOHBMETRC-UHFFFAOYSA-N | |
11182279 | |
CC1=CC(=CC(=C1)P(C2=CC(=CC(=C2)C)C)C3=CC(=NC(=C3C4=C(N=C(C=C4P(C5=CC(=CC(=C5)C)C)C6=CC(=CC(=C6)C)C)OC)OC)OC)OC)C |
C46H50N2O4P2 | |
MFCD04974235 | |
unii-1a4eo1493y, unii-n2pcw56063, r-4,4'-bis bis 3,5-dimethylphenyl phosphino-2,2',6,6'-tetramethoxy-3,3'-bipyridine, s---2,2',6,6'-tetramethoxy-4,4'-bis di 3,5-xylyl phosphino-3,3'-bipyridine, s-4,4'-bis bis 3,5-dimethylphenyl phosphino-2,2',6,6'-tetramethoxy-3,3'-bipyridine, xyl-p-phos, r, xyl-p-phos, s, r-xyl-p-phos mi, s-xyl-p-phos mi, xyl-p-phos | |
[3-[4-bis(3,5-dimethylphenyl)phosphanyl-2,6-dimethoxypyridin-3-yl]-2,6-dimethoxypyridin-4-yl]-bis(3,5-dimethylphenyl)phosphane |
Safety and Handling
TSCA : No
storageNote1 :
- Packaged under Argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only