Please login to your online account to display your discounted pricing

(S)-2-(1-BOC-2-pyrrolidinyl)acetic acid, 97%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC399970010

 View more versions of this product

Catalog No. AC399970010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥96.0% (GC)
Formula Weight 229.27
Quantity 1g
Color White to off-white
Melting Point 97° to 101°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material (S)-2-(1-BOC-2-pyrrolidinyl)acetic acid
NMR Consistent with structure (1H-NMR)
Assay Percent Range 97%
Specific Rotation −36.6° to −40.6° (20°C, 589nm) (c=1, 4 in DMF)
Molecular Formula C11H19NO4
CAS 56502-01-3
MDL Number MFCD01862939
Synonym BOC-L-beta-homoproline
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (S)-2-(1-BOC-2-pyrrolidinyl)acetic acid
CAS 56502-01-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C11H19NO4
Formula Weight 229.27
MDL Number MFCD01862939
Synonym BOC-L-beta-homoproline
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.