Please login to your online account to display your discounted pricing

S 18986, Tocris™

Positive allosteric modulator of AMPA

$239.63 - $968.50

Chemical Identifiers

Chemical Name or Material S 18986
Assay Percent Range >98%
Molecular Formula C10H12N2O2S
Formula Weight 224.28
Synonym 2,3,3a,4-Tetrahydro-1H-pyrrolo[2,1-c][1,2,4]benzothiadiazine-5,5-dioxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

391110

 
r & d systems
RDS391110
10mg Each for $239.63

391150

 
r & d systems
RDS391150
50mg Each for $968.50
Description & Specifications

Specifications

Formula Weight 224.28
Quantity 10mg
Molecular Formula C10H12N2O2S
Synonym 2,3,3a,4-Tetrahydro-1H-pyrrolo[2,1-c][1,2,4]benzothiadiazine-5,5-dioxide
Recommended Storage Store at -20°C
CAS 175340-20-2
Chemical Name or Material S 18986
Assay Percent Range >98%
Safety and Handling

Store at -20°C