Please login to your online account to display your discounted pricing

S 14506 hydrochloride, Tocris

Highly potent 5-HT1A agonist; displays unique binding mechanism

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material S 14506 hydrochloride
Assay Percent Range >99%
Molecular Formula C24H26FN3O2·HCl
Formula Weight 443.94
Synonym 4-Fluoro-N-[2-[4-(7-methoxy-1-naphthalenyl)-1-piperazinyl]ethyl]benzamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

177110

 
r & d systems
177110
10mg Each for $244.81

177150

 
r & d systems
177150
50mg Each for $997.23
Description & Specifications

Specifications

Formula Weight 443.94
Quantity 10mg
Chemical Name or Material S 14506 hydrochloride
Assay Percent Range >99%
Molecular Formula C24H26FN3O2·HCl
Recommended Storage Desiccate at Room Temperature
CAS 286369-38-8
Synonym 4-Fluoro-N-[2-[4-(7-methoxy-1-naphthalenyl)-1-piperazinyl]ethyl]benzamide hydrochloride
Safety and Handling

Desiccate at Room Temperature