missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-1-(tert-Butoxycarbonyl)-2-cyanopyrrolidine, TCI America™

$33.61 - $103.89

Chemical Identifiers

CAS 228244-04-0
Molecular Formula C10H16N2O2
Molecular Weight (g/mol) 196.25
MDL Number MFCD01861224
InChI Key MDMSZBHMBCNYNO-QMMMGPOBSA-N
Synonym s-1-boc-2-cyanopyrrolidine, s-tert-butyl 2-cyanopyrrolidine-1-carboxylate, s-1-n-boc-2-cyano-pyrrolidine, tert-butyl 2s-2-cyanopyrrolidine-1-carboxylate, s-1-boc-2-cyano-pyrrolidine, s-1-n-boc-2-cyanopyrrolidine, s-1-boc-2-pyrrolidinecarbonitrile, s-n-boc-pyrrolidine-2-carbonitrile, s---1-boc-2-cyanopyrrolidine, s-n-boc-2-pyrrolidinonitrile
PubChem CID 6951263
IUPAC Name tert-butyl (2S)-2-cyanopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B48781G View Documents TCI America
B48781G
1g Each for $33.61
 
B48785G View Documents TCI America
B48785G
5g Each for $103.89
 
Specifications

Chemical Identifiers

228244-04-0
196.25
MDMSZBHMBCNYNO-QMMMGPOBSA-N
6951263
CC(C)(C)OC(=O)N1CCCC1C#N
C10H16N2O2
MFCD01861224
s-1-boc-2-cyanopyrrolidine, s-tert-butyl 2-cyanopyrrolidine-1-carboxylate, s-1-n-boc-2-cyano-pyrrolidine, tert-butyl 2s-2-cyanopyrrolidine-1-carboxylate, s-1-boc-2-cyano-pyrrolidine, s-1-n-boc-2-cyanopyrrolidine, s-1-boc-2-pyrrolidinecarbonitrile, s-n-boc-pyrrolidine-2-carbonitrile, s---1-boc-2-cyanopyrrolidine, s-n-boc-2-pyrrolidinonitrile
tert-butyl (2S)-2-cyanopyrrolidine-1-carboxylate

Specifications

White-Yellow
1g
C10H16N2O2
s-1-boc-2-cyanopyrrolidine, s-tert-butyl 2-cyanopyrrolidine-1-carboxylate, s-1-n-boc-2-cyano-pyrrolidine, tert-butyl 2s-2-cyanopyrrolidine-1-carboxylate, s-1-boc-2-cyano-pyrrolidine, s-1-n-boc-2-cyanopyrrolidine, s-1-boc-2-pyrrolidinecarbonitrile, s-n-boc-pyrrolidine-2-carbonitrile, s---1-boc-2-cyanopyrrolidine, s-n-boc-2-pyrrolidinonitrile
CC(C)(C)OC(=O)N1CCCC1C#N
196.25
196.25
≥96.0% (GC)
33°C
228244-04-0
MFCD01861224
MDMSZBHMBCNYNO-QMMMGPOBSA-N
tert-butyl (2S)-2-cyanopyrrolidine-1-carboxylate
6951263
Crystal-Lump at 20°C
(S)-1-(tert-Butoxycarbonyl)-2-cyanopyrrolidine
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator