missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-1-Phenylpropylamine, TCI America™

Manufacturer:  TCI America P150825G

 View more versions of this product

Catalog No. P150825G



Specifications

Specifications

(S)-(-)-1-Phenylpropylamine
-69°C
3789-59-1
MFCD00082356
s-1-phenylpropan-1-amine, pubchem6025, s---1-amino-1-phenylpropane, 1s-1-phenylpropan-1-amine, s---1-phenylpropylamine, s-1-phenylpropylamine, s---alpha-ethylbenzylamine, s---methylbenzylamine, 1-phenyl-propylamine, s---, a-ethylbenzylamine
CCC(C1=CC=CC=C1)N
135.21
135.21
≥98.0% (GC)
Yellow
25g
C9H13N
2735
AQFLVLHRZFLDDV-VIFPVBQESA-N
(1S)-1-phenylpropan-1-amine
6993773
Clear Liquid at 20°C

Chemical Identifiers

3789-59-1
135.21
AQFLVLHRZFLDDV-VIFPVBQESA-N
6993773
CCC(C1=CC=CC=C1)N
C9H13N
MFCD00082356
s-1-phenylpropan-1-amine, pubchem6025, s---1-amino-1-phenylpropane, 1s-1-phenylpropan-1-amine, s---1-phenylpropylamine, s-1-phenylpropylamine, s---alpha-ethylbenzylamine, s---methylbenzylamine, 1-phenyl-propylamine, s---, a-ethylbenzylamine
(1S)-1-phenylpropan-1-amine
Safety and Handling

Safety and Handling

TSCA : No

Documents