Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-1-Phenylpropylamine, ChiPros 99+%, ee 99%

Chemical Identifiers

Chemical Name or Material (S)-(-)-1-Phenylpropylamine
CAS 3789-59-1
Molecular Formula C9H13N
Formula Weight 135.21
MDL Number MFCD00082356
UN Number UN2735
Beilstein 2412016
Synonym (S)-(-)-1-Amino-1-phenylpropane; (S)-(-)-alpha-Ethylbenzylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1632003

  SDS
alfa aesar™
L1632003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1632014

  SDS
alfa aesar™
L1632014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1632006

  SDS
alfa aesar™
L1632006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 135.21
CAS 3789-59-1
Molecular Formula C9H13N
Chemical Name or Material (S)-(-)-1-Phenylpropylamine
Health Hazard 2 GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
Synonym (S)-(-)-1-Amino-1-phenylpropane; (S)-(-)-alpha-Ethylbenzylamine
Beilstein 2412016
Boiling Point 205°C
UN Number UN2735
Assay ≥99%
Assay Percent Notes ee 99%
MDL Number MFCD00082356
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Quantity 1g
Health Hazard 3 GHS P Statement
P280-P273-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Health Hazard 1 Danger
Flash Point 77°C (170°F)
Odor Amine-like
Melting Point −69°C
Optical Rotation −20° (Neat)
Safety and Handling

Danger

GHS P Statement
P280-P273-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.