Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-1-Phenylpropyl isocyanate, 95%

Chemical Identifiers

Chemical Name or Material (S)-(-)-1-Phenylpropyl isocyanate
CAS 164033-12-9
Molecular Formula C10H11NO
Formula Weight 161.2
MDL Number MFCD05664090
UN Number UN2206
Beilstein 6799586
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2024603

  SDS
alfa aesar™
L2024603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 161.2
Chemical Name or Material (S)-(-)-1-Phenylpropyl isocyanate
Assay 95%
Molecular Formula C10H11NO
MDL Number MFCD05664090
CAS 164033-12-9
UN Number UN2206
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Optical Rotation −38.5° (c=1, Chloroform)
Beilstein 6799586
Health Hazard 1 Danger
Boiling Point 105°C (17mmHg)
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C