missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-1-Phenylethyl isocyanate, 98%

Manufacturer:  Alfa Aesar™ L1114106

 View more versions of this product

Catalog No. AAL1114106



Description

Description

Applications
(S)-(-)-1-Phenylethyl isocyanate act by inhibition of cell proliferation and induction of apoptosis. It helps to prevent lung cancer and esophageal cancer. It is a fine chemical intermediate.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to moisture. Recommended storage temperature is 2 - 8°C.
Specifications

Specifications

(S)-(-)-1-Phenylethyl isocyanate
Pungent
55°C to 56°C (2.5mmHg)
14649-03-7
MFCD00002036
4230972
JJSCUXAFAJEQGB-QMMMGPOBSA-N
[(1S)-1-isocyanatoethyl]benzene
2724173
98%
65°C (149°F)
−10.5° (Neat)
5g
C9H9NO
UN2206
s---1-phenylethyl isocyanate, 1s-1-isocyanatoethyl benzene, s---alpha-methylbenzyl isocyanate, benzene, 1s-1-isocyanatoethyl, isocyanic acid s---a-methylbenzyl ester, s-alpha-methylbenzyl isocyanate, s---1-phenylethylisocyanate, isocyanic acid s---alpha-methylbenzyl ester, isocyanic acid s---methylbenzyl ester, 1-isocyanatoethyl benzene #
CC(C1=CC=CC=C1)N=C=O
147.177
147.18

Chemical Identifiers

14649-03-7
147.177
JJSCUXAFAJEQGB-QMMMGPOBSA-N
2724173
CC(C1=CC=CC=C1)N=C=O
C9H9NO
MFCD00002036
s---1-phenylethyl isocyanate, 1s-1-isocyanatoethyl benzene, s---alpha-methylbenzyl isocyanate, benzene, 1s-1-isocyanatoethyl, isocyanic acid s---a-methylbenzyl ester, s-alpha-methylbenzyl isocyanate, s---1-phenylethylisocyanate, isocyanic acid s---alpha-methylbenzyl ester, isocyanic acid s---methylbenzyl ester, 1-isocyanatoethyl benzene #
[(1S)-1-isocyanatoethyl]benzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents