Please login to your online account to display your discounted pricing

(S)-1-Phenyl-1,3-propanediol, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  417760010

 View more versions of this product

Catalog No. AC417760010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 152.19
Physical Form Crystalline Powder and Chunks
Quantity 1g
Color White
Melting Point 62° to 66°C
Specific Rotation Condition −54.50° (22°C c=1,1,2-dichloroethane)
CAS Max % 100.0
CAS 96854-34-1
Infrared Spectrum Authentic
Chemical Name or Material (S)-1-Phenyl-1, 3-propanediol
Assay Percent Range 98%
Molecular Formula C9H12O2
CAS Min % 97.5
MDL Number MFCD00221510
Packaging Glass bottle
Specific Rotation -53° to -56° (22°C, 589nm) (c=1, 1,2-dichloroethane)

Specifications

Chemical Name or Material (S)-1-Phenyl-1, 3-propanediol
CAS 96854-34-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H12O2
Formula Weight 152.19
MDL Number MFCD00221510