Please login to your online account to display your discounted pricing

(S)-(+)-1-Phenyl-1,2-ethanediol, ≥99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-(+)-1-Phenyl-1, 2-ethanediol
CAS 25779-13-9
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C8H10O2
Formula Weight 138.16
MDL Number MFCD00066256
Synonym (S)-(+)-α, β-Dihydroxyethylbenzene
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC264640010

  SDS
thermo fisher scientific
264640010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Flakes
Formula Weight 138.16
Quantity 1g
Color White
Specific Rotation Condition +59.5° (20°C c=2,1,2-dichloroEt)
Melting Point 65° to 68°C
Merck Index 14, 8861
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material (S)-(+)-1-Phenyl-1, 2-ethanediol
Synonym (S)-(+)-α, β-Dihydroxyethylbenzene
Packaging Glass bottle
Assay Percent Range 99+%
Molecular Formula C8H10O2
CAS 25779-13-9
Specific Rotation +58.5° to +60.5° (20°C, 589nm) (c=2, 1,2-dichloroethane)
MDL Number MFCD00066256
Infrared Spectrum Authentic