Learn More
S-(1-Oxido-2-pyridyl)-N,N,N',N'-tetramethylthiuronium tetrafluoroborate, 98+%, Thermo Scientific™
$59.14 - $59.14
Chemical Identifiers
| CAS | 255825-38-8 |
|---|---|
| Molecular Formula | C10H16BF4N3OS |
| Molecular Weight (g/mol) | 313.12 |
| MDL Number | MFCD05664718 |
| InChI Key | LHLFXDQURZVFLK-UHFFFAOYSA-N |
| Synonym | tott, 2-1-oxy-pyridin-2-yl-1,1,3,3-tetramethylisothiouronium tetrafluoroborate, 2-dimethylamino dimethyliminio methyl sulfanyl pyridin-1-ium-1-olate tetrafluoroborate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiouronium tetrafluoroborate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium tetrafluoroborate, s-1-oxido-2-pyridyl-n,n,n',n'-tetramethylthiuronium tetrafluoroborate, s-2-pyridyl-n,n,n',n'-tetramethylthiuronium n-oxide tetrafluoroborate, s-1-oxido-2-pyridyl-n,n,n',n'-tetramethylthiuronium tetrafluoroborate nmr, tott, 2-1-oxy-pyridin-2-yl-1,1,3,3-tetramethylisothiouronium tetrafluoroborate, dimethylamino-1-oxidopyridin-1-ium-2-yl sulfanyl-methylene-dimethyl-ammonium tetrafluoroborate |
| PubChem CID | 10543173 |
| IUPAC Name | [dimethylamino-(1-oxidopyridin-1-ium-2-yl)sulfanylmethylidene]-dimethylazanium;tetrafluoroborate |
| SMILES | F[B-](F)(F)F.CN(C)C(SC1=CC=CC=[N+]1[O-])=[N+](C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5440403
|
Thermo Scientific Chemicals
H5440403 |
1 g |
Each for $59.14
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 255825-38-8 | |
| 313.12 | |
| LHLFXDQURZVFLK-UHFFFAOYSA-N | |
| 10543173 | |
| F[B-](F)(F)F.CN(C)C(SC1=CC=CC=[N+]1[O-])=[N+](C)C |
| C10H16BF4N3OS | |
| MFCD05664718 | |
| tott, 2-1-oxy-pyridin-2-yl-1,1,3,3-tetramethylisothiouronium tetrafluoroborate, 2-dimethylamino dimethyliminio methyl sulfanyl pyridin-1-ium-1-olate tetrafluoroborate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiouronium tetrafluoroborate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium tetrafluoroborate, s-1-oxido-2-pyridyl-n,n,n',n'-tetramethylthiuronium tetrafluoroborate, s-2-pyridyl-n,n,n',n'-tetramethylthiuronium n-oxide tetrafluoroborate, s-1-oxido-2-pyridyl-n,n,n',n'-tetramethylthiuronium tetrafluoroborate nmr, tott, 2-1-oxy-pyridin-2-yl-1,1,3,3-tetramethylisothiouronium tetrafluoroborate, dimethylamino-1-oxidopyridin-1-ium-2-yl sulfanyl-methylene-dimethyl-ammonium tetrafluoroborate | |
| [dimethylamino-(1-oxidopyridin-1-ium-2-yl)sulfanylmethylidene]-dimethylazanium;tetrafluoroborate |
Specifications
| 255825-38-8 | |
| C10H16BF4N3OS | |
| 1 g | |
| tott, 2-1-oxy-pyridin-2-yl-1,1,3,3-tetramethylisothiouronium tetrafluoroborate, 2-dimethylamino dimethyliminio methyl sulfanyl pyridin-1-ium-1-olate tetrafluoroborate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiouronium tetrafluoroborate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium tetrafluoroborate, s-1-oxido-2-pyridyl-n,n,n',n'-tetramethylthiuronium tetrafluoroborate, s-2-pyridyl-n,n,n',n'-tetramethylthiuronium n-oxide tetrafluoroborate, s-1-oxido-2-pyridyl-n,n,n',n'-tetramethylthiuronium tetrafluoroborate nmr, tott, 2-1-oxy-pyridin-2-yl-1,1,3,3-tetramethylisothiouronium tetrafluoroborate, dimethylamino-1-oxidopyridin-1-ium-2-yl sulfanyl-methylene-dimethyl-ammonium tetrafluoroborate | |
| F[B-](F)(F)F.CN(C)C(SC1=CC=CC=[N+]1[O-])=[N+](C)C | |
| 313.12 | |
| 314.13 | |
| S-(1-Oxido-2-pyridyl)-N,N,N',N'-tetramethylthiuronium tetrafluoroborate |
| 111°C to 116°C | |
| MFCD05664718 | |
| Moisture sensitive | |
| LHLFXDQURZVFLK-UHFFFAOYSA-N | |
| [dimethylamino-(1-oxidopyridin-1-ium-2-yl)sulfanylmethylidene]-dimethylazanium;tetrafluoroborate | |
| 10543173 | |
| ≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Store at -20°C; Store under Nitrogen