missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ S-(1-Oxido-2-pyridyl)-N,N,N',N'-tetramethylthiuronium tetrafluoroborate, 98+%

Manufacturer:  Alfa Aesar™ H5440403

 View more versions of this product

Catalog No. AAH5440403



Specifications

Specifications

S-(1-Oxido-2-pyridyl)-N,N,N',N'-tetramethylthiuronium tetrafluoroborate
1g
C10H16BF4N3OS
2-1-oxy-pyridin-2-yl-1,1,3,3-tetramethylisothiouronium tetrafluoroborate
[B-](F)(F)(F)F.CN(C)C(=[N+](C)C)SC1=CC=CC=[N+]1[O-]
313.122
314.13
111°C to 116°C
255825-38-8
MFCD05664718
LHLFXDQURZVFLK-UHFFFAOYSA-N
[dimethylamino-(1-oxidopyridin-1-ium-2-yl)sulfanylmethylidene]-dimethylazanium;tetrafluoroborate
10543173
≥98%

Chemical Identifiers

255825-38-8
313.122
LHLFXDQURZVFLK-UHFFFAOYSA-N
10543173
[B-](F)(F)(F)F.CN(C)C(=[N+](C)C)SC1=CC=CC=[N+]1[O-]
C10H16BF4N3OS
MFCD05664718
2-1-oxy-pyridin-2-yl-1,1,3,3-tetramethylisothiouronium tetrafluoroborate
[dimethylamino-(1-oxidopyridin-1-ium-2-yl)sulfanylmethylidene]-dimethylazanium;tetrafluoroborate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : −30° to −10°C

Documents