missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ S-(1-Oxido-2-pyridyl)-N,N,N',N'-tetramethylthiuronium hexafluorophosphate, 98+%

$27.47 - $454.84

Chemical Identifiers

CAS 212333-72-7
Molecular Formula C10H16F6N3OPS
Molecular Weight (g/mol) 371.282
MDL Number MFCD05865266
InChI Key UCOGEMMJHLHOAW-UHFFFAOYSA-N
Synonym acmc-209fij
PubChem CID 10571248
IUPAC Name [dimethylamino-(1-oxidopyridin-1-ium-2-yl)sulfanylmethylidene]-dimethylazanium;hexafluorophosphate
SMILES CN(C)C(=[N+](C)C)SC1=CC=CC=[N+]1[O-].F[P-](F)(F)(F)(F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5427703 View Documents Alfa Aesar™
H5427703
1g Each for $27.47
 
AAH5427706 View Documents Alfa Aesar™
H5427706
5g Each for $109.28
 
AAH5427714 View Documents Alfa Aesar™
H5427714
25g Each for $454.84
 
Specifications

Chemical Identifiers

212333-72-7
371.282
UCOGEMMJHLHOAW-UHFFFAOYSA-N
10571248
CN(C)C(=[N+](C)C)SC1=CC=CC=[N+]1[O-].F[P-](F)(F)(F)(F)F
C10H16F6N3OPS
MFCD05865266
acmc-209fij
[dimethylamino-(1-oxidopyridin-1-ium-2-yl)sulfanylmethylidene]-dimethylazanium;hexafluorophosphate

Specifications

Moisture sensitive
1g
C10H16F6N3OPS
acmc-209fij
CN(C)C(=[N+](C)C)SC1=CC=CC=[N+]1[O-].F[P-](F)(F)(F)(F)F
371.282
371.28
S-(1-Oxido-2-pyridyl)-N,N,N',N'-tetramethylthiuronium hexafluorophosphate
120°C to 125°C
212333-72-7
MFCD05865266
UCOGEMMJHLHOAW-UHFFFAOYSA-N
[dimethylamino-(1-oxidopyridin-1-ium-2-yl)sulfanylmethylidene]-dimethylazanium;hexafluorophosphate
10571248
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Store at -20°C; Store under Nitrogen