missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-1-Octyn-3-ol 99.0+%, TCI America™

$284.17 - $864.73

Chemical Identifiers

CAS 32556-71-1
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00191475
InChI Key VUGRNZHKYVHZSN-MRVPVSSYSA-N
Synonym s-1-octyn-3-ol, s---1-octyn-3-ol, s-3-hydroxy-1-octyne, 1-octyn-3-ol, 3s, 3s-1-octyn-3-ol, unii-d8ho63xx92, 3s-oct-1-yn-3-ol, 3s-hydroxy-1-octyne, --1-octyn-3-ol, 1-octyn-3-ol, s
PubChem CID 2734534
IUPAC Name (3S)-oct-1-yn-3-ol
SMILES CCCCCC(C#C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
O02351G View Documents TCI America
O02351G
1g Each for $284.17
 
O02355G View Documents TCI America
O02355G
5g Each for $864.73
 
Specifications

Chemical Identifiers

32556-71-1
126.199
VUGRNZHKYVHZSN-MRVPVSSYSA-N
2734534
CCCCCC(C#C)O
C8H14O
MFCD00191475
s-1-octyn-3-ol, s---1-octyn-3-ol, s-3-hydroxy-1-octyne, 1-octyn-3-ol, 3s, 3s-1-octyn-3-ol, unii-d8ho63xx92, 3s-oct-1-yn-3-ol, 3s-hydroxy-1-octyne, --1-octyn-3-ol, 1-octyn-3-ol, s
(3S)-oct-1-yn-3-ol

Specifications

130°C
1g
C8H14O
s-1-octyn-3-ol, s---1-octyn-3-ol, s-3-hydroxy-1-octyne, 1-octyn-3-ol, 3s, 3s-1-octyn-3-ol, unii-d8ho63xx92, 3s-oct-1-yn-3-ol, 3s-hydroxy-1-octyne, --1-octyn-3-ol, 1-octyn-3-ol, s
CCCCCC(C#C)O
126.199
126.20
≥99.0% (GC)
Yellow
32556-71-1
MFCD00191475
VUGRNZHKYVHZSN-MRVPVSSYSA-N
(3S)-oct-1-yn-3-ol
2734534
Clear Liquid at 20°C
(S)-1-Octyn-3-ol [omega Side-Chain Unit for PG Synthesis]
Safety and Handling

Safety and Handling

TSCA : No