missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-1-Octyn-3-ol 99.0+%, TCI America™

Manufacturer:  TCI America O02351G

 View more versions of this product

Catalog No. O02351G



Specifications

Specifications

(S)-1-Octyn-3-ol [omega Side-Chain Unit for PG Synthesis]
Yellow
32556-71-1
MFCD00191475
VUGRNZHKYVHZSN-MRVPVSSYSA-N
(3S)-oct-1-yn-3-ol
2734534
Clear Liquid at 20°C
130°C
1g
C8H14O
s-1-octyn-3-ol, s---1-octyn-3-ol, s-3-hydroxy-1-octyne, 1-octyn-3-ol, 3s, 3s-1-octyn-3-ol, unii-d8ho63xx92, 3s-oct-1-yn-3-ol, 3s-hydroxy-1-octyne, --1-octyn-3-ol, 1-octyn-3-ol, s
CCCCCC(C#C)O
126.199
126.20
≥99.0% (GC)

Chemical Identifiers

32556-71-1
126.199
VUGRNZHKYVHZSN-MRVPVSSYSA-N
2734534
CCCCCC(C#C)O
C8H14O
MFCD00191475
s-1-octyn-3-ol, s---1-octyn-3-ol, s-3-hydroxy-1-octyne, 1-octyn-3-ol, 3s, 3s-1-octyn-3-ol, unii-d8ho63xx92, 3s-oct-1-yn-3-ol, 3s-hydroxy-1-octyne, --1-octyn-3-ol, 1-octyn-3-ol, s
(3S)-oct-1-yn-3-ol
Safety and Handling

Safety and Handling

TSCA : No

Documents