Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-1-Octyn-3-ol, 98+%

$58.18 - $158.54

Chemical Identifiers

Chemical Name or Material (S)-(-)-1-Octyn-3-ol
CAS 32556-71-1
Molecular Formula C8H14O
Formula Weight 126.2
MDL Number MFCD00191475
Beilstein 2037703
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1904603

 
alfa aesar™
L1904603
1g Each for $158.54

AAL19046MD

 
alfa aesar™
L19046MD
250mg Each for $58.18
Description & Specifications

Specifications

Formula Weight 126.2
CAS 32556-71-1
Chemical Name or Material (S)-(-)-1-Octyn-3-ol
Molecular Formula C8H14O
MDL Number MFCD00191475
Beilstein 2037703
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H227
Toxic if swallowed.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P301+P310-P405-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 63°C (145°F)
Optical Rotation −9.9° (Neat)
Assay ≥98%
Boiling Point 100°C (20mmHg)
Quantity 1g

Applications
It is a key intermediate for the synthesis of prostaglandins. It is used in the synthesis of (R)-3-aminooctanoic acid.

Notes
Store away from strong oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.

Safety and Handling

Danger

GHS P Statement
P210-P280-P301+P310-P405-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H227
Toxic if swallowed.
Combustible liquid.