Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-1-Methyl-3-hydroxypyrrolidine, 98%, ee 99%

$115.72 - $320.95

Chemical Identifiers

Chemical Name or Material (S)-(+)-1-Methyl-3-hydroxypyrrolidine
CAS 104641-59-0
Molecular Formula C5H11NO
Formula Weight 101.15
MDL Number MFCD03788748
Beilstein 4349394
Synonym (S)-(+)-1-Methyl-3-pyrrolidinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1969503

  SDS
alfa aesar™
L1969503
1g Each for $320.95

AAL19695MD

  SDS
alfa aesar™
L19695MD
250mg Each for $115.72
Description & Specifications

Specifications

Formula Weight 101.15
Chemical Name or Material (S)-(+)-1-Methyl-3-hydroxypyrrolidine
Assay Percent Notes ee 99%
MDL Number MFCD03788748
CAS 104641-59-0
Synonym (S)-(+)-1-Methyl-3-pyrrolidinol
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Flash Point 70°C (158°F)
Beilstein 4349394
Boiling Point 103° to 106°C (35mmHg)
Assay 98%
Molecular Formula C5H11NO
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Safety and Handling

Warning

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.