missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-1-Methoxy-2-propylamine, ChiPros 98+%, ee 96+%

$37.01 - $37.01

Chemical Identifiers

CAS 99636-32-5
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD01075730
InChI Key NXMXETCTWNXSFG-BYPYZUCNSA-N
Synonym s-+-1-methoxy-2-propylamine, 2-propanamine, 1-methoxy-, 2s, 2s-1-methoxypropan-2-amine, s-2-amino-1-methoxypropane, s-1-methoxypropan-2-amine, s-1-methoxy-2-propylamine, s-1methoxy-2-aminopropane, s-1-methoxy-2-propanamine, s-1-methoxy-2-aminopropane, s-2-amino-3-methoxypropane
PubChem CID 7021536
IUPAC Name (2S)-1-methoxypropan-2-amine
SMILES CC(COC)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1632503 View Documents Alfa Aesar™
L1632503
1g Each for $37.01
 
Description

Description

Applications
(S)-(+)-1-Methoxy-2-propylamine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Fully miscible in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. It is sensitive to air. Store under dry inert gas. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

99636-32-5
89.138
NXMXETCTWNXSFG-BYPYZUCNSA-N
7021536
CC(COC)N
C4H11NO
MFCD01075730
s-+-1-methoxy-2-propylamine, 2-propanamine, 1-methoxy-, 2s, 2s-1-methoxypropan-2-amine, s-2-amino-1-methoxypropane, s-1-methoxypropan-2-amine, s-1-methoxy-2-propylamine, s-1methoxy-2-aminopropane, s-1-methoxy-2-propanamine, s-1-methoxy-2-aminopropane, s-2-amino-3-methoxypropane
(2S)-1-methoxypropan-2-amine

Specifications

9°C (48°F)
98°C to 99°C
1g
C4H11NO
UN2733
s-+-1-methoxy-2-propylamine, 2-propanamine, 1-methoxy-, 2s, 2s-1-methoxypropan-2-amine, s-2-amino-1-methoxypropane, s-1-methoxypropan-2-amine, s-1-methoxy-2-propylamine, s-1methoxy-2-aminopropane, s-1-methoxy-2-propanamine, s-1-methoxy-2-aminopropane, s-2-amino-3-methoxypropane
CC(COC)N
89.138
89.14
(S)-(+)-1-Methoxy-2-propylamine
+11.5° (Neat)
-95°C
99636-32-5
MFCD01075730
5725728
NXMXETCTWNXSFG-BYPYZUCNSA-N
(2S)-1-methoxypropan-2-amine
7021536
≥98%
ee ≥96%
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318-H302
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P210-P280-P273-P305+P351+P338-P309-P310
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

TSCA : TSCA