Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-1-Fmoc-4-oxopiperidine-2-carboxylic acid, 97%

Chemical Identifiers

Chemical Name or Material (S)-(-)-1-Fmoc-4-oxopiperidine-2-carboxylic acid
CAS 1221793-43-6
Formula Weight 365.38
MDL Number MFCD03094868
Synonym Fmoc-L-Homopro(4-oxo)-OH; (S)-(-)-1-Fmoc-4-oxohomoproline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6642803

 
alfa aesar™
H6642803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66428MD

 
alfa aesar™
H66428MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 228° to 223°C
Formula Weight 365.38
Chemical Name or Material (S)-(-)-1-Fmoc-4-oxopiperidine-2-carboxylic acid
MDL Number MFCD03094868
Assay 97%
CAS 1221793-43-6
Synonym Fmoc-L-Homopro(4-oxo)-OH; (S)-(-)-1-Fmoc-4-oxohomoproline
Physical Form Powder
Sensitivity Moisture Sensitive
Quantity 1g
Color White to yellow
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C