missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-(-)-1-Ethyl-2-pyrrolidinecarboxamide 98.0+%, TCI America™
$300.29 - $1013.65
Chemical Identifiers
CAS | 114812-34-9 |
---|---|
Molecular Formula | C7H14N2O |
Molecular Weight (g/mol) | 142.202 |
MDL Number | MFCD00191361 |
InChI Key | LQOASEHNECNLNM-LURJTMIESA-N |
Synonym | N-Ethyl-L-prolinamide |
PubChem CID | 10866443 |
IUPAC Name | (2S)-1-ethylpyrrolidine-2-carboxamide |
SMILES | CCN1CCCC1C(=O)N |
Chemical Identifiers
114812-34-9 | |
142.202 | |
LQOASEHNECNLNM-LURJTMIESA-N | |
10866443 | |
CCN1CCCC1C(=O)N |
C7H14N2O | |
MFCD00191361 | |
N-Ethyl-L-prolinamide | |
(2S)-1-ethylpyrrolidine-2-carboxamide |
Specifications
114812-34-9 | |
White-Yellow | |
MFCD00191361 | |
N-Ethyl-L-prolinamide | |
CCN1CCCC1C(=O)N | |
142.202 | |
142.20 | |
Crystalline Powder |
110°C | |
C7H14N2O | |
1 g | |
LQOASEHNECNLNM-LURJTMIESA-N | |
(2S)-1-ethylpyrrolidine-2-carboxamide | |
10866443 | |
≥98.0% (GC,T) | |
(S)-(-)-1-Ethyl-2-pyrrolidinecarboxamide |
Safety and Handling
TSCA : No