Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-1-Cyclohexylethylamine, ChiPros 98%, ee 97+%

Chemical Identifiers

Chemical Name or Material (S)-(+)-1-Cyclohexylethylamine
CAS 17430-98-7
Molecular Formula C8H17N
Formula Weight 127.23
MDL Number MFCD00066261
UN Number UN2733
Beilstein 2935069
Synonym (S)-(+)-alpha-Methylcyclohexanemethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1906603

  SDS
alfa aesar™
L1906603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1906606

  SDS
alfa aesar™
L1906606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1906614

  SDS
alfa aesar™
L1906614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 127.23
Assay 98%
Assay Percent Notes ee ≥97%
MDL Number MFCD00066261
CAS 17430-98-7
Synonym (S)-(+)-alpha-Methylcyclohexanemethylamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Flash Point 51°C (123°F)
Chemical Name or Material (S)-(+)-1-Cyclohexylethylamine
Molecular Formula C8H17N
UN Number UN2733
Beilstein 2935069
pH 11
Boiling Point 177° to 178°C
Quantity 1g
Odor Amine-like
Optical Rotation +4° (Neat)
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.