missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-1-Boc-4,4-diethoxypiperidine-2-carboxylic acid, 98%

$150.39 - $447.93

Chemical Identifiers

CAS 661458-35-1
Molecular Formula C15H27NO6
Formula Weight 317.38
MDL Number MFCD08056311
Synonym (S)-1-(tert-Butoxycarbonyl)-4,4-diethoxypiperidine-2-carboxylic Acid
Chemical Name or Material (S)-1-Boc-4,4-diethoxypiperidine-2-carboxylic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH25905MC SDS Alfa Aesar™
H25905MC
100mg Each for $150.39
AAH25905ME SDS Alfa Aesar™
H25905ME
500mg Each for $447.93
Specifications Description & Specifications

Chemical Identifiers

661458-35-1
317.38
(S)-1-(tert-Butoxycarbonyl)-4,4-diethoxypiperidine-2-carboxylic Acid
C15H27NO6
MFCD08056311
(S)-1-Boc-4,4-diethoxypiperidine-2-carboxylic acid

Specifications

661458-35-1
317.38
(S)-1-(tert-Butoxycarbonyl)-4,4-diethoxypiperidine-2-carboxylic Acid
(S)-1-Boc-4,4-diethoxypiperidine-2-carboxylic acid
C15H27NO6
MFCD08056311
100mg
98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C