Please login to your online account to display your discounted pricing

(S)-1-BOC-3-methylpiperazine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-1-BOC-3-Methylpiperazine
CAS 147081-29-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C10H20N2O2
Formula Weight 200.28
MDL Number MFCD02683204
Synonym tert-Butyl (S)-3-methyl-1-piperazinecarboxylate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC429700050

  SDS
thermo fisher scientific
AC429700050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC429700010

 
thermo fisher scientific
AC429700010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥97.5% (GC)
Formula Weight 200.28
Quantity 5g
Color White to yellow
Melting Point 45° to 49°C
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym tert-Butyl (S)-3-methyl-1-piperazinecarboxylate
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material (S)-1-BOC-3-Methylpiperazine
Assay Percent Range 98%
Molecular Formula C10H20N2O2
CAS 147081-29-6
CAS Min % 97.5
MDL Number MFCD02683204
Specific Rotation −23° to −27° (20°C, 589nm) (c=2.5, MeOH)
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.