Please login to your online account to display your discounted pricing

(S)-1-BOC-3-methylpiperazine, 98%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC429700010

 View more versions of this product

Catalog No. AC429700010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥97.5% (GC)
Formula Weight 200.28
Quantity 1g
Color White to yellow
Melting Point 45° to 49°C
CAS Max % 100.0
Specific Rotation −23° to −27° (20°C, 589nm) (c=2.5, MeOH)
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 98%
Molecular Formula C10H20N2O2
CAS Min % 97.5
Chemical Name or Material (S)-1-BOC-3-Methylpiperazine
CAS 147081-29-6
MDL Number MFCD02683204
Synonym tert-Butyl (S)-3-methyl-1-piperazinecarboxylate
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material (S)-1-BOC-3-Methylpiperazine
CAS 147081-29-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C10H20N2O2
Formula Weight 200.28
MDL Number MFCD02683204
Synonym tert-Butyl (S)-3-methyl-1-piperazinecarboxylate
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.