missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-(+)-1-Boc-3-fluoropyrrolidine, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6688903
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
(S)-(+)-1-Boc-3-fluoropyrrolidine | |
Colorless to Yellow | |
74°C to 80°C (2 mmHg) | |
C9H16FNO2 | |
MFCD04114064 | |
UN2810 | |
s-1-boc-3-fluoropyrrolidine, s-n-boc-3-fluoropyrrolidine, s-tert-butyl 3-fluoropyrrolidine-1-carboxylate, tert-butyl 3s-3-fluoropyrrolidine-1-carboxylate, 1-pyrrolidinecarboxylic acid, 3-fluoro-, 1,1-dimethylethyl ester, 3s, n-trans-boc-3s-fluoropyrrolidine, tert-butyl s-3-fluoropyrrolidine-1-carboxylate, 3s-n-boc-3-fluoropyrrolidine, s-+-1-boc-3-fluoropyrrolidine | |
CC(C)(C)OC(=O)N1CCC(C1)F | |
189.23 | |
189.23 | |
Liquid |
479253-00-4 | |
1.069 g/mL | |
103°C (217°F) | |
1.441 | |
1 g | |
Moisture sensitive | |
SUECTKVSIDXQQE-ZETCQYMHSA-N | |
tert-butyl (3S)-3-fluoropyrrolidine-1-carboxylate | |
18453464 | |
96% |
Chemical Identifiers
479253-00-4 | |
189.23 | |
SUECTKVSIDXQQE-ZETCQYMHSA-N | |
18453464 | |
CC(C)(C)OC(=O)N1CCC(C1)F |
C9H16FNO2 | |
MFCD04114064 | |
s-1-boc-3-fluoropyrrolidine, s-n-boc-3-fluoropyrrolidine, s-tert-butyl 3-fluoropyrrolidine-1-carboxylate, tert-butyl 3s-3-fluoropyrrolidine-1-carboxylate, 1-pyrrolidinecarboxylic acid, 3-fluoro-, 1,1-dimethylethyl ester, 3s, n-trans-boc-3s-fluoropyrrolidine, tert-butyl s-3-fluoropyrrolidine-1-carboxylate, 3s-n-boc-3-fluoropyrrolidine, s-+-1-boc-3-fluoropyrrolidine | |
tert-butyl (3S)-3-fluoropyrrolidine-1-carboxylate |
Safety and Handling
GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
P264b-P270-P280i-P301+P310-P305+P351+P338-P330-P501c
H301-H319
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen