Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-1-Boc-3-aminopyrrolidine, 98%, ee 99%

Chemical Identifiers

Chemical Name or Material (S)-(-)-1-Boc-3-aminopyrrolidine
CAS 147081-44-5
Molecular Formula C9H18N2O2
Formula Weight 186.25
MDL Number MFCD03419271
UN Number UN2735
Beilstein 8312125
Synonym (S)-1-(tert-Butoxycarbonyl)-3-aminopyrrolidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1969603

  SDS
alfa aesar™
L1969603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1969606

  SDS
alfa aesar™
L1969606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 186.25
Chemical Name or Material (S)-(-)-1-Boc-3-aminopyrrolidine
MDL Number MFCD03419271
CAS 147081-44-5
Health Hazard 3 GHS P Statement
P260-P280-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Health Hazard 1 Danger
UN Number UN2735
Assay 98%
Synonym (S)-1-(tert-Butoxycarbonyl)-3-aminopyrrolidine
Quantity 1g
Recommended Storage 2° to 8°C
Beilstein 8312125
Assay Percent Notes ee 99%
Optical Rotation −3° (Neat)
Molecular Formula C9H18N2O2
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Boiling Point 115°C (4mmHg)
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

Applications
(S)-(-)-1-Boc-3-aminopyrrolidine is specifically used to protect amine in the solid phase synthesis of peptides. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Notes
Store under inert gas. It is sensitive to air. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P280-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

2° to 8°C