Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-1-Boc-3-(aminomethyl)pyrrolidine, 97%

Chemical Identifiers

Chemical Name or Material (S)-(+)-1-Boc-3-(aminomethyl)pyrrolidine
CAS 199175-10-5
Formula Weight 200.28
MDL Number MFCD02179397
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6635903

 
alfa aesar™
H6635903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66359MD

 
alfa aesar™
H66359MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 200.28
Chemical Name or Material (S)-(+)-1-Boc-3-(aminomethyl)pyrrolidine
MDL Number MFCD02179397
CAS 199175-10-5
Physical Form Liquid
Assay 97%
Refractive Index 1.4780
Sensitivity Air Sensitive
Quantity 1g
Color Colorless to yellow
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C