missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-1-Benzyloxycarbonyl-5-oxopyrrolidine-2-carboxylic acid, 98%

$79.11 - $314.59

Chemical Identifiers

CAS 32159-21-0
Molecular Formula C13H13NO5
Molecular Weight (g/mol) 263.249
MDL Number MFCD00037352
InChI Key VHSFUGXCSGOKJX-JTQLQIEISA-N
Synonym z-pyr-oh, cbz-l-pyroglutamic acid, z-l-pyroglutamic acid, benzyloxycarbonyl-l-pyroglutamic acid, s-5-oxo-pyrrolidine-1,2-dicarboxylic acid 1-benzyl ester, s-1-benzyloxy carbonyl-5-oxopyrrolidine-2-carboxylic acid, s-1-benzyloxycarbonyl-5-oxopyrrolidine-2-carboxylic acid, s-1-benzyloxycarbonyl-5-oxopyrrolidine-2-carboxylic acid, 2s-5-oxo-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid, z-pglu-oh
PubChem CID 637648
IUPAC Name (2S)-5-oxo-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
SMILES C1CC(=O)N(C1C(=O)O)C(=O)OCC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6288606 View Documents Alfa Aesar™
H6288606
5g Each for $79.11
 
AAH6288614 View Documents Alfa Aesar™
H6288614
25g Each for $314.59
 
Specifications

Chemical Identifiers

32159-21-0
263.249
VHSFUGXCSGOKJX-JTQLQIEISA-N
637648
C1CC(=O)N(C1C(=O)O)C(=O)OCC2=CC=CC=C2
C13H13NO5
MFCD00037352
z-pyr-oh, cbz-l-pyroglutamic acid, z-l-pyroglutamic acid, benzyloxycarbonyl-l-pyroglutamic acid, s-5-oxo-pyrrolidine-1,2-dicarboxylic acid 1-benzyl ester, s-1-benzyloxy carbonyl-5-oxopyrrolidine-2-carboxylic acid, s-1-benzyloxycarbonyl-5-oxopyrrolidine-2-carboxylic acid, s-1-benzyloxycarbonyl-5-oxopyrrolidine-2-carboxylic acid, 2s-5-oxo-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid, z-pglu-oh
(2S)-5-oxo-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid

Specifications

5g
C13H13NO5
z-pyr-oh, cbz-l-pyroglutamic acid, z-l-pyroglutamic acid, benzyloxycarbonyl-l-pyroglutamic acid, s-5-oxo-pyrrolidine-1,2-dicarboxylic acid 1-benzyl ester, s-1-benzyloxy carbonyl-5-oxopyrrolidine-2-carboxylic acid, s-1-benzyloxycarbonyl-5-oxopyrrolidine-2-carboxylic acid, s-1-benzyloxycarbonyl-5-oxopyrrolidine-2-carboxylic acid, 2s-5-oxo-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid, z-pglu-oh
C1CC(=O)N(C1C(=O)O)C(=O)OCC2=CC=CC=C2
263.249
263.25
(S)-1-Benzyloxycarbonyl-5-oxopyrrolidine-2-carboxylic acid
32159-21-0
MFCD00037352
VHSFUGXCSGOKJX-JTQLQIEISA-N
(2S)-5-oxo-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
637648
98%
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C