missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-1-Benzyl-3-pyrrolidinol, TCI America™

$116.11 - $403.34

Chemical Identifiers

CAS 101385-90-4
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD00075485
InChI Key YQMXOIAIYXXXEE-NSHDSACASA-N
Synonym s-1-benzylpyrrolidin-3-ol, s-1-benzyl-3-pyrrolidinol, s-1-benzyl-3-hydroxypyrrolidine, 3s-1-benzylpyrrolidin-3-ol, s-3-hydroxy-1-benzylpyrrolidine, s-1-benzyl-pyrrolidin-3-ol, s---1-benzyl-3-pyrrolidinol, s-1-n-benzyl-3-hydroxypyrrolidine, s-bhp, s-n-benzyl-3-hydroxypyrrolidine
PubChem CID 2733875
IUPAC Name (3S)-1-benzylpyrrolidin-3-ol
SMILES C1CN(CC1O)CC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B15335G View Documents TCI America
B15335G
5g Each for $116.11
 
B153325G View Documents TCI America
B153325G
25g Each for $403.34
 
Specifications

Chemical Identifiers

101385-90-4
177.247
YQMXOIAIYXXXEE-NSHDSACASA-N
2733875
C1CN(CC1O)CC2=CC=CC=C2
C11H15NO
MFCD00075485
s-1-benzylpyrrolidin-3-ol, s-1-benzyl-3-pyrrolidinol, s-1-benzyl-3-hydroxypyrrolidine, 3s-1-benzylpyrrolidin-3-ol, s-3-hydroxy-1-benzylpyrrolidine, s-1-benzyl-pyrrolidin-3-ol, s---1-benzyl-3-pyrrolidinol, s-1-n-benzyl-3-hydroxypyrrolidine, s-bhp, s-n-benzyl-3-hydroxypyrrolidine
(3S)-1-benzylpyrrolidin-3-ol

Specifications

120°C
5g
C11H15NO
s-1-benzylpyrrolidin-3-ol, s-1-benzyl-3-pyrrolidinol, s-1-benzyl-3-hydroxypyrrolidine, 3s-1-benzylpyrrolidin-3-ol, s-3-hydroxy-1-benzylpyrrolidine, s-1-benzyl-pyrrolidin-3-ol, s---1-benzyl-3-pyrrolidinol, s-1-n-benzyl-3-hydroxypyrrolidine, s-bhp, s-n-benzyl-3-hydroxypyrrolidine
C1CN(CC1O)CC2=CC=CC=C2
177.247
177.25
≥98.0% (GC)
Yellow
101385-90-4
MFCD00075485
YQMXOIAIYXXXEE-NSHDSACASA-N
(3S)-1-benzylpyrrolidin-3-ol
2733875
Clear Liquid at 20°C
(S)-1-Benzyl-3-pyrrolidinol
Safety and Handling

Safety and Handling

TSCA : No