Please login to your online account to display your discounted pricing

(S)-(-)-1-Benzyl-3-pyrrolidinol, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC398360010

 View more versions of this product

Catalog No. AC398360010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 177.25
Physical Form Liquid
Quantity 1g
Color Clear light yellow to pale brown
Density 1.07g/mL
CAS Max % 100.0
Refractive Index 1.547 to 1.549 (20°C, 589nm)
Specific Rotation -3.0° to -4.5° (20°C, 589nm) (C=5, methanol)
Molecular Formula C11H15NO
Assay Percent Range 98%
Chemical Name or Material (S)-(-)-1-Benzyl-3-pyrrolidinol
CAS 101385-90-4
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
CAS Min % 97.5
MDL Number MFCD00075485
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (S)-(-)-1-Benzyl-3-pyrrolidinol
CAS 101385-90-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H15NO
Formula Weight 177.25
MDL Number MFCD00075485
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents