missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-1-Benzyl-3-pyrrolidinol, 99%, ACROS Organics™

Chemical Identifiers

CAS 101385-90-4
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C11H15NO
Formula Weight 177.25
MDL Number MFCD00075485
Chemical Name or Material (S)-(-)-1-Benzyl-3-pyrrolidinol
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC398360050 SDS Thermo Fisher Scientific
AC398360050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC398360010 SDS Thermo Fisher Scientific
AC398360010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

101385-90-4
100.0
C11H15NO
MFCD00075485
≥97.5%
97.5
98%
177.25
(S)-(-)-1-Benzyl-3-pyrrolidinol

Specifications

177.25
5g
1.07g/mL
Authentic
Glass bottle
C11H15NO
97.5
-3.0° to -4.5° (20°C, 589nm) (C=5, methanol)
≥97.5%
Liquid
Clear light yellow to pale brown
100.0
1.547 to 1.549 (20°C, 589nm)
101385-90-4
98%
MFCD00075485
(S)-(-)-1-Benzyl-3-pyrrolidinol

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.