Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-1-Benzyl-3-hydroxypyrrolidine, 99%, ee 99%

$48.64 - $151.93

Chemical Identifiers

Chemical Name or Material (S)-(-)-1-Benzyl-3-hydroxypyrrolidine
CAS 101385-90-4
Molecular Formula C11H15NO
Formula Weight 177.25
MDL Number MFCD00075485
Beilstein 4982831
Synonym (S)-(-)-1-Benzyl-3-pyrrolidinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1949703

 
alfa aesar™
L1949703
1g Each for $48.64

AAL1949706

 
alfa aesar™
L1949706
5g Each for $151.93
Description & Specifications

Specifications

Formula Weight 177.25
Molecular Formula C11H15NO
Chemical Name or Material (S)-(-)-1-Benzyl-3-hydroxypyrrolidine
Assay 99%
Assay Percent Notes ee 99%
MDL Number MFCD00075485
CAS 101385-90-4
Synonym (S)-(-)-1-Benzyl-3-pyrrolidinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Boiling Point 116°C (0.9mmHg)
Quantity 1g
Flash Point 148°C (298°F)
Optical Rotation −3.7° (c=5 in Methanol)
Beilstein 4982831
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.