Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-1-Benzyl-3-hydroxypiperidine, 97%

$102.36 - $260.14

Chemical Identifiers

Chemical Name or Material (S)-(+)-1-Benzyl-3-hydroxypiperidine
CAS 91599-79-0
Molecular Formula C12H17NO
Formula Weight 191.27
MDL Number MFCD03426359
Synonym (S)-1-Benzyl-3-piperidinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3061503

 
alfa aesar™
H3061503
1g Each for $260.14

AAH30615MD

 
alfa aesar™
H30615MD
250mg Each for $102.36
Description & Specifications

Specifications

Formula Weight 191.27
Chemical Name or Material (S)-(+)-1-Benzyl-3-hydroxypiperidine
Assay 97%
Molecular Formula C12H17NO
MDL Number MFCD03426359
CAS 91599-79-0
Synonym (S)-1-Benzyl-3-piperidinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Recommended Storage 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C