Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-1-Benzyl-3-aminopyrrolidine, 99%

Chemical Identifiers

Chemical Name or Material (S)-(+)-1-Benzyl-3-aminopyrrolidine
CAS 114715-38-7
Molecular Formula C11H16N2
Formula Weight 176.26
MDL Number MFCD00082766
UN Number UN2735
Beilstein 6477593
Synonym (3S)-(+)-3-Amino-1-benzylpyrrolidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0944903

 
alfa aesar™
L0944903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0944906

 
alfa aesar™
L0944906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 176.26
Chemical Name or Material (S)-(+)-1-Benzyl-3-aminopyrrolidine
CAS 114715-38-7
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Assay 99%
Molecular Formula C11H16N2
MDL Number MFCD00082766
UN Number UN2735
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Quantity 1g
Flash Point 140°C (284°F)
Beilstein 6477593
Synonym (3S)-(+)-3-Amino-1-benzylpyrrolidine
Boiling Point 100° to 105°C (3mmHg)
Optical Rotation +5° (c=10 in Ethanol)
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.