Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-1-Benzyl-3-acetamidopyrrolidine, 98%, ee 99%

$204.92 - $204.92

Chemical Identifiers

Chemical Name or Material (S)-(-)-1-Benzyl-3-acetamidopyrrolidine
CAS 114636-30-5
Molecular Formula C13H18N2O
Formula Weight 218.3
MDL Number MFCD03788746
Beilstein 6479772
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1968906

  SDS
alfa aesar™
L1968906
5g Each for $204.92
Description & Specifications

Specifications

Formula Weight 218.3
Chemical Name or Material (S)-(-)-1-Benzyl-3-acetamidopyrrolidine
Assay 98%
Molecular Formula C13H18N2O
Assay Percent Notes ee 99%
MDL Number MFCD03788746
CAS 114636-30-5
Beilstein 6479772
Melting Point 44° to 46°C
Quantity 5g
Optical Rotation −38° (c=1 in Methanol)
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C