Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-1-Aminoindane, ChiPros 99+%, ee 99+%

$120.20 - $443.31

Chemical Identifiers

Chemical Name or Material (S)-(+)-1-Aminoindane
CAS 61341-86-4
Molecular Formula C9H11N
Formula Weight 133.19
MDL Number MFCD00216670
Beilstein 2206708
Synonym (S)-(+)-1-Indanamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1930303

  SDS
alfa aesar™
L1930303
1g Each for $120.20

AAL1930306

  SDS
alfa aesar™
L1930306
5g Each for $443.31
Description & Specifications

Specifications

Formula Weight 133.19
Chemical Name or Material (S)-(+)-1-Aminoindane
Assay ≥99%
Assay Percent Notes ee ≥99%
Molecular Formula C9H11N
MDL Number MFCD00216670
CAS 61341-86-4
Beilstein 2206708
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Boiling Point 225°C
Quantity 1g
Flash Point 94°C (201°F)
Synonym (S)-(+)-1-Indanamine
Optical Rotation +16.5° (c=1.5 in Methanol)

Applications
(S)-(+)-1-Aminoindane is the S-enantiomer of 1-aminoindane, which is the metabolite of the monoamine oxidase-B inhibitor rasagiline. It is also used as a chiral and organic building block.

Solubility
Miscible with toluene. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and carbondioxide

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.