missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(+)-1-Amino-2-propanol, +98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 218980010

 View more versions of this product

Catalog No. AC218980010


 

Specifications Description & Specifications

Specifications

(S)-(+)-1-Amino-2-propanol
Authentic
100.0
1.4465 to 1.4495 (20°C, 589nm)
Clear colorless to pale yellow
1g
98+%
CH3CH(OH)CH2NH2
MFCD00064429
(S)-(+)-Isopropanolamine
2799-17-9
98.0
Glass bottle
+16° to +20° (20°C, 589nm) (c=1.8, H2O)
Liquid After Melting
≥98.0%
C3H9NO
75.11
04, IV, 1665

Chemical Identifiers

2799-17-9
100.0
C3H9NO
75.11
(S)-(+)-1-Amino-2-propanol
98.0
98+%
CH3CH(OH)CH2NH2
MFCD00064429
≥98.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger