Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-1-Amino-2-propanol, 98%

$76.65 - $76.65

Chemical Identifiers

Chemical Name or Material (S)-(+)-1-Amino-2-propanol
CAS 2799-17-9
Molecular Formula C3H9NO
Formula Weight 75.11
MDL Number MFCD00064429
UN Number UN3259
Beilstein 1718868
Synonym (S)-(+)-Isopropanolamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1410203

 
alfa aesar™
L1410203
1g Each for $76.65

AAL14102MD

 
alfa aesar™
L14102MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 75.11
Chemical Name or Material (S)-(+)-1-Amino-2-propanol
Assay 98%
Molecular Formula C3H9NO
MDL Number MFCD00064429
CAS 2799-17-9
UN Number UN3259
Beilstein 1718868
Synonym (S)-(+)-Isopropanolamine
Health Hazard 2 GHS H Statement
H314-H318-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 24° to 26°C
Quantity 1g
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
RTECS Number UA5775000
Health Hazard 1 Danger
Boiling Point 160°C
Flash Point 71°C (159°F)
Optical Rotation +18.5° (c=1.8 in Water)
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.