Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-1-(4-Fluorophenyl)ethyl isocyanate, 95%

Chemical Identifiers

Chemical Name or Material (S)-(-)-1-(4-Fluorophenyl)ethyl isocyanate
CAS 745783-74-8
Molecular Formula C9H8FNO
Formula Weight 165.17
MDL Number MFCD05664076
UN Number UN2206
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2023103

  SDS
alfa aesar™
L2023103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 165.17
Chemical Name or Material (S)-(-)-1-(4-Fluorophenyl)ethyl isocyanate
MDL Number MFCD05664076
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 95%
Molecular Formula C9H8FNO
CAS 745783-74-8
UN Number UN2206
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C