missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-1-(4-Chlorophenyl)ethylamine, ChiPros™ 97%, ee 98%

$163.93 - $604.31

Chemical Identifiers

CAS 4187-56-8
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.625
MDL Number MFCD00671640
InChI Key PINPOEWMCLFRRB-LURJTMIESA-N
Synonym 1s-1-4-chlorophenyl ethan-1-amine
PubChem CID 1550852
IUPAC Name (1S)-1-(4-chlorophenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)Cl)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1905206 View Documents Alfa Aesar™
L1905206
5g Each for $163.93
 
AAL1905214 View Documents Alfa Aesar™
L1905214
25g Each for $604.31
 
Description

Description

Applications
It is used as a substrate in palladium catalysts on alkaline-earth supports for racemization and dynamic kinetic resolution of benzylic amines.

Solubility
Soluble in water at 25°C 11 g/L.

Notes
Air sensitive. Store away from acids, air, acid chlorides, acid anhydrides and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications

Chemical Identifiers

4187-56-8
155.625
PINPOEWMCLFRRB-LURJTMIESA-N
1550852
CC(C1=CC=C(C=C1)Cl)N
C8H10ClN
MFCD00671640
1s-1-4-chlorophenyl ethan-1-amine
(1S)-1-(4-chlorophenyl)ethanamine

Specifications

115°C (239°F)
−31° (Neat)
5g
C8H10ClN
UN2922
1s-1-4-chlorophenyl ethan-1-amine
CC(C1=CC=C(C=C1)Cl)N
155.625
155.63
(S)-(-)-1-(4-Chlorophenyl)ethylamine
Pungent
232°C
4187-56-8
MFCD00671640
2412316
PINPOEWMCLFRRB-LURJTMIESA-N
(1S)-1-(4-chlorophenyl)ethanamine
1550852
97%
ee 98%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H330-H314-H318
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.