missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-1-(4-Chlorophenyl)ethylamine, ChiPros™ 97%, ee 98%

Manufacturer:  Alfa Aesar™ L1905206

 View more versions of this product

Catalog No. AAL1905206



Description

Description

Applications
It is used as a substrate in palladium catalysts on alkaline-earth supports for racemization and dynamic kinetic resolution of benzylic amines.

Solubility
Soluble in water at 25°C 11 g/L.

Notes
Air sensitive. Store away from acids, air, acid chlorides, acid anhydrides and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications

Specifications

(S)-(-)-1-(4-Chlorophenyl)ethylamine
Pungent
232°C
4187-56-8
C8H10ClN
UN2922
1s-1-4-chlorophenyl ethan-1-amine, 1s-1-4-chlorophenyl ethanamine, 1s-1-4-chlorophenyl ethylamine, a-methylbenzylamine, pubchem15221, s---1-4-chlorophenyl ethylamine, s-1-4-chloro-phenyl ethylamine, s-1-4-chlorophenyl ethanamine, s-1-4-chlorophenyl ethylamine, s-4-chloro-, s-4-chloro-alpha-methylbenzylamine
PINPOEWMCLFRRB-LURJTMIESA-N
(1S)-1-(4-chlorophenyl)ethanamine
1550852
97%
1.5418
115°C (239°F)
−31° (Neat)
5g
ee 98%
MFCD00671640
2412316
Soluble in water at 25°C 11g/L.
CC(C1=CC=C(C=C1)Cl)N
155.625
155.63
1.11
Air Sensitive

Chemical Identifiers

4187-56-8
155.625
PINPOEWMCLFRRB-LURJTMIESA-N
1550852
CC(C1=CC=C(C=C1)Cl)N
C8H10ClN
MFCD00671640
1s-1-4-chlorophenyl ethan-1-amine, 1s-1-4-chlorophenyl ethanamine, 1s-1-4-chlorophenyl ethylamine, a-methylbenzylamine, pubchem15221, s---1-4-chlorophenyl ethylamine, s-1-4-chloro-phenyl ethylamine, s-1-4-chlorophenyl ethanamine, s-1-4-chlorophenyl ethylamine, s-4-chloro-, s-4-chloro-alpha-methylbenzylamine
(1S)-1-(4-chlorophenyl)ethanamine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H330-H314-H318
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P272-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P330-P333+P313-P362-P501c

H302-H315-H317-H318-H330

DOTInformation : Hazard Class: 6.1; Packaging Group: II

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

Documents