missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-1-(4-Chlorophenyl)ethylamine, ChiPros™ 94%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6671503

 View more versions of this product

Catalog No. AAH6671503



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(S)-(-)-1-(4-Chlorophenyl)ethylamine
4187-56-8
1.110g/mL
1 g
Pungent
1.5418
MFCD00671640
2412316
PINPOEWMCLFRRB-LURJTMIESA-N
(1S)-1-(4-chlorophenyl)ethanamine
1550852
94%
ChiPros™
Yellow
232°C
115°C (239°F)
Liquid
C8H10ClN
UN2922
s-1-4-chlorophenyl ethylamine, s-1-4-chloro-phenyl ethylamine, 1s-1-4-chlorophenyl ethanamine, 1s-1-4-chlorophenyl ethanamine, s-4-chloro-alpha-methylbenzylamine, s---1-4-chlorophenyl ethylamine, pubchem15221, pubchem15221, a-methylbenzylamine, 1s-1-4-chlorophenyl ethylamine, 1s-1-4-chlorophenyl ethylamine
CC(C1=CC=C(C=C1)Cl)N
155.625
155.63

Chemical Identifiers

4187-56-8
155.625
PINPOEWMCLFRRB-LURJTMIESA-N
1550852
CC(C1=CC=C(C=C1)Cl)N
C8H10ClN
MFCD00671640
s-1-4-chlorophenyl ethylamine, s-1-4-chloro-phenyl ethylamine, 1s-1-4-chlorophenyl ethanamine, 1s-1-4-chlorophenyl ethanamine, s-4-chloro-alpha-methylbenzylamine, s---1-4-chlorophenyl ethylamine, pubchem15221, pubchem15221, a-methylbenzylamine, 1s-1-4-chlorophenyl ethylamine, 1s-1-4-chlorophenyl ethylamine
(1S)-1-(4-chlorophenyl)ethanamine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H330-H314-H318
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P272-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P330-P333+P313-P362-P501c

H302-H315-H317-H318-H330

DOTInformation : Hazard Class: 6.1; Packaging Group: II

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only