missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-1-(4-Chlorophenyl)ethylamine, 95+%, (sum of isomers), 95% e.e., ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC444110250

 View more versions of this product

Catalog No. AC444110250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(S)-1-(4-Chlorophenyl)ethylamine
115°C
1.5410 to 1.5430
− 23.50
25g
4187-56-8
C8H10ClN
1s-1-4-chlorophenyl ethan-1-amine, 1s-1-4-chlorophenyl ethanamine, 1s-1-4-chlorophenyl ethylamine, a-methylbenzylamine, pubchem15221, s---1-4-chlorophenyl ethylamine, s-1-4-chloro-phenyl ethylamine, s-1-4-chlorophenyl ethanamine, s-1-4-chlorophenyl ethylamine, s-4-chloro-, s-4-chloro-alpha-methylbenzylamine
PINPOEWMCLFRRB-LURJTMIESA-N
(1S)-1-(4-chlorophenyl)ethanamine
1550852
95+%
1.1150g/mL
Glass bottle
1.115
232.0°C
95% e.e
95% min. (GC)
ClC6H4CH(CH3)NH2
Solubility in water: moderately soluble
CC(C1=CC=C(C=C1)Cl)N
155.63
155.63

Chemical Identifiers

4187-56-8
155.63
1s-1-4-chlorophenyl ethan-1-amine, 1s-1-4-chlorophenyl ethanamine, 1s-1-4-chlorophenyl ethylamine, a-methylbenzylamine, pubchem15221, s---1-4-chlorophenyl ethylamine, s-1-4-chloro-phenyl ethylamine, s-1-4-chlorophenyl ethanamine, s-1-4-chlorophenyl ethylamine, s-4-chloro-, s-4-chloro-alpha-methylbenzylamine
(1S)-1-(4-chlorophenyl)ethanamine
C8H10ClN
PINPOEWMCLFRRB-LURJTMIESA-N
1550852
CC(C1=CC=C(C=C1)Cl)N
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye damage.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

SDS Documents